missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GLYCTK Partial ORF (NP_660305.2, 332 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00132158-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16101827

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described. [provided by RefSeq]

Sequence: VLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRV
Especificaciones

Especificaciones

NP_660305.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRV
RUO
GLYCTK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
132158
GLYCTK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GLYCTK1/HBEBP2/HBEBP4/HBeAgBP4A
GLYCTK
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales