Learn More
Abnova™ Human GFM1 Partial ORF (NP_079272, 653 a.a. - 751 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00085476-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. [provided by RefSeq]
Sequence: EVVAPNEFQGQVIAGINRRHGVITGQDGVEDYFTLYADVPLNDMFGYSTELRSCTEGKGEYTMEYSRYQPCLPSTQEDVINKYLEATGQLPVKKGKAKNEspecificaciones
NP_079272 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EVVAPNEFQGQVIAGINRRHGVITGQDGVEDYFTLYADVPLNDMFGYSTELRSCTEGKGEYTMEYSRYQPCLPSTQEDVINKYLEATGQLPVKKGKAKN | |
RUO | |
GFM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
85476 | |
GFM1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COXPD1/EFG/EFG1/EFGM/EGF1/FLJ12662/FLJ13632/FLJ20773/GFM/hEFG1 | |
GFM1 | |
Recombinant | |
wheat germ expression system |