missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GFI1B Partial ORF (NP_004179, 2 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008328-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: PRSFLVKSKMAHTYHQPRVQEDEPLWPPALTPVPRDQAPSNSPVLSTLFPNQCLDWTNLKREPELEQDQNLARMAPAPEGPIVLSRPQDGDSPLSDSPPFYKPSFSWDTEspecificaciones
NP_004179 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PRSFLVKSKMAHTYHQPRVQEDEPLWPPALTPVPRDQAPSNSPVLSTLFPNQCLDWTNLKREPELEQDQNLARMAPAPEGPIVLSRPQDGDSPLSDSPPFYKPSFSWDT | |
RUO | |
GFI1B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8328 | |
GFI1B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GFI1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |