missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GATA5 Full-length ORF (NP_536721.1, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00140628-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108223

  • 341.70€ / 10 microgramos
Envío estimado: 24-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a transcription factor that contains two GATA-type zinc fingers. The encoded protein is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha), and this interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. In other organisms, similar proteins may be involved in the establishment of cardiac smooth muscle cell diversity. [provided by RefSeq]

Sequence: MYQSLALAASPRQAAYADSGSFLHAPGAGSPMFVPPARVPSMLSYLSGCEPSPQPPELAARPGWAQTATADSSAFGPGSPHPPAAHPPGATAFPFAHSPSGPGSGGSAGGRDGSAYQGALLPREQFAAPLGRPVGTSYSATYPAYVSPDVAQSWTAGPFDGSVLHGLPGRRPTFVSDFLEEFPGEGRECVNCGALSTPLWRRDGTGHYLCNACGLYHKMNGVNRPLVRPQKRLSSSRRAGLCCTNCHTTNTTLWRRNSEGEPVCNACGLYMKLHGVPRPLAMKKESIQTRKRKPKTIAKARGSSGSTRNASASPSAVASTDSSAATSKAKPSLASPVCPGPSMAPQASGQEDDSLAPGHLEFKFEPEDFAFPSTAPSPQAGLRGALRQEAWCALALA
Especificaciones

Especificaciones

NP_536721.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
bB379O24.1
GATA5
Yes
wheat germ expression system
Liquid
140628
GATA5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYQSLALAASPRQAAYADSGSFLHAPGAGSPMFVPPARVPSMLSYLSGCEPSPQPPELAARPGWAQTATADSSAFGPGSPHPPAAHPPGATAFPFAHSPSGPGSGGSAGGRDGSAYQGALLPREQFAAPLGRPVGTSYSATYPAYVSPDVAQSWTAGPFDGSVLHGLPGRRPTFVSDFLEEFPGEGRECVNCGALSTPLWRRDGTGHYLCNACGLYHKMNGVNRPLVRPQKRLSSSRRAGLCCTNCHTTNTTLWRRNSEGEPVCNACGLYMKLHGVPRPLAMKKESIQTRKRKPKTIAKARGSSGSTRNASASPSAVASTDSSAATSKAKPSLASPVCPGPSMAPQASGQEDDSLAPGHLEFKFEPEDFAFPSTAPSPQAGLRGALRQEAWCALALA
RUO
GATA5
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales