Learn More
Abnova™ Human FOXA1 Full-length ORF (NP_004487.2, 1 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003169-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. [provided by RefSeq]
Sequence: MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYANPGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSPMAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQNQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGSGSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELKTPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAYEQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTSEspecificaciones
NP_004487.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HNF3A/MGC33105/TCF3A | |
FOXA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3169 | |
FOXA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYANPGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSPMAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQNQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGSGSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELKTPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAYEQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTS | |
RUO | |
FOXA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |