Learn More
ACROBiosystems Human Flt-3 / Flk-2 Protein, Fc Tag
362.89€ - 2661.24€
Especificaciones
Número de acceso | NP_004110.2 |
---|---|
Peso molecular | 84.8 kDa |
Fuente | Human Flt-3, Fc Tag (FL3-H5258) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1). |
Inmunógeno | Asn 27 - Ser 543 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16477539
|
ACROBiosystems
FL3-H5258-100UG |
100 μg |
362.89€
100 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16487539
|
ACROBiosystems
FL3-H5258-1MG |
1 mg |
2661.24€
1 miligramo |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Flt-3(Receptor-type tyrosine-protein kinase FLT3) is also known as FLK-2(Fetal liver kinase-2), STK-1(Stem cell tyrosine kinase 1), CD135. FLT3 is a cytokine receptor which belongs to the receptor tyrosine kinase class III. Tyrosine-protein kinase that acts as cell-surface receptor for the cytokine FLT3LG and regulates differentiation, proliferation and survival of hematopoietic progenitor cells and of dendritic cells. Promotes phosphorylation of SHC1 and AKT1, and activation of the downstream effector MTOR. Promotes activation of RAS signaling and phosphorylation of downstream kinases, including MAPK1/ERK2 and/or MAPK3/ERK1. Mutations that cause constitutive kinase activity promote cell proliferation and resistance to apoptosis via the activation of multiple signaling pathways.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.8 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_004110.2 | |
Human Flt-3, Fc Tag (FL3-H5258) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1). | |
-20°C | |
Flt-3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
84.8 kDa | |
Asn 27 - Ser 543 | |
Flt-3,Flk-2,STK-1,CD135,FLK2,FLT-3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |