missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLJ20097 Partial ORF (NP_060137, 862 a.a. - 964 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055610-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: GYANVKKCSNEGRALMQLDFQQFLMKLEKLTDIRPIPDKEFVETYIKAYYLTENDMERWIKEHREYSTKQLTNLVNVCLGSHINKKARQKLLAAIDDIDRPKREspecificaciones
NP_060137 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GYANVKKCSNEGRALMQLDFQQFLMKLEKLTDIRPIPDKEFVETYIKAYYLTENDMERWIKEHREYSTKQLTNLVNVCLGSHINKKARQKLLAAIDDIDRPKR | |
RUO | |
CCDC132 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55610 | |
FLJ20097 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20097/FLJ23581/KIAA1861/MGC176659 | |
CCDC132 | |
Recombinant | |
wheat germ expression system |