missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLJ10769 Full-length ORF (AAH41028.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055739-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MVTRAGAGTAVAGAVVVALLSAALALYGPPLDAVLERAFSLRKAHSIKDMENTLQLVRNIIPPLSSTKHKGQDGRIGVVGGCQEYTGAPYFAAISALKVGADLSHVFCASAAAPVIKAYSPELIVHPVLDSPNAVHEVEKWLPRLHALVVGPGLGRDDALLRNVQGILEVSKARDIPVVIDADGLWLVAQQPALIHGYRKAVLTPNHVEFSRLYDAVLRGPMDSDDSHGSVLRLSQALGNVTVVQKGERDILSNGQQVLVCSQEGSSRRCGGQGDLLSGSLGVLVHWALLAGPQKTNGSSPLLVAAFGACSLTRQCNHQAFQKHGRSTTTSDMIAEVGAAFSKLFETEspecificaciones
AAH41028.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10769/FLJ34548/LP3298 | |
CARKD | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55739 | |
FLJ10769 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVTRAGAGTAVAGAVVVALLSAALALYGPPLDAVLERAFSLRKAHSIKDMENTLQLVRNIIPPLSSTKHKGQDGRIGVVGGCQEYTGAPYFAAISALKVGADLSHVFCASAAAPVIKAYSPELIVHPVLDSPNAVHEVEKWLPRLHALVVGPGLGRDDALLRNVQGILEVSKARDIPVVIDADGLWLVAQQPALIHGYRKAVLTPNHVEFSRLYDAVLRGPMDSDDSHGSVLRLSQALGNVTVVQKGERDILSNGQQVLVCSQEGSSRRCGGQGDLLSGSLGVLVHWALLAGPQKTNGSSPLLVAAFGACSLTRQCNHQAFQKHGRSTTTSDMIAEVGAAFSKLFET | |
RUO | |
CARKD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |