Learn More
Abnova™ Human FIG4 Full-length ORF (NP_055660.1, 1 a.a. - 907 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009896-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene belongs to the SAC domain-containing protein gene family. The SAC domain, approximately 400 amino acids in length and consisting of seven conserved motifs, has been shown to possess phosphoinositide phosphatase activity. The yeast homolog, Sac1p, is involved in the regulation of various phosphoinositides, and affects diverse cellular functions such as actin cytoskeleton organization, Golgi function, and maintenance of vacuole morphology. Membrane-bound phosphoinositides function as signaling molecules and play a key role in vesicle trafficking in eukaryotic cells. Mutations in this gene have been associated with Charcot-Marie-Tooth disease, type 4J. [provided by RefSeq]
Sequence: MPTAAAPIISSVQKLVLYETRARYFLVGSNNAETKYRVLKIDRTEPKDLVIIDDRHVYTQQEVRELLGRLDLGNRTKMGQKGSSGLFRAVSAFGVVGFVRFLEGYYIVLITKRRKMADIGGHAIYKVEDTNMIYIPNDSVRVTHPDEARYLRIFQNVDLSSNFYFSYSYDLSHSLQYNLTVLRMPLEMLKSEMTQNRQESFDIFEDEGLITQGGSGVFGICSEPYMKYVWNGELLDIIKSTVHRDWLLYIIHGFCGQSKLLIYGRPVYVTLIARRSSKFAGTRFLKRGANCEGDVANEVETEQILCDASVMSFTAGSYSSYVQVRGSVPLYWSQDISTMMPKPPITLDQADPFAHVAALHFDQMFQRFGSPIIILNLVKEREKRKHERILSEELVAAVTYLNQFLPPEHTIVYIPWDMAKYTKSKLCNVLDRLNVIAESVVKKTGFFVNRPDSYCSILRPDEKWNELGGCVIPTGRLQTGILRTNCVDCLDRTNTAQFMVGKCALAYQLYSLGLIDKPNLQFDTDAVRLFEELYEDHGDTLSLQYGGSQLVHRVKTYRKIAPWTQHSKDIMQTLSRYYSNAFSDADRQDSINLFLGVFHPTEGKPHLWELPTDFYLHHKNTMRLLPTRRSYTYWWTPEVIKHLPLPYDEVICAVNLKKLIVKKFHKYEEEIDIHNEFFRPYELSSFDDTFCLAMTSSARDFMPKTVGIDPSPFTVRKPDETGKSVLGNKSNREEAVLQRKTAASAPPPPSEEAVSSSSEDDSGTDREEEGSVSQRSTPVKMTDAGDSAKVTENVVQPMKELYGINLSDGLSEEDFSIYSRFVQLGQSQHKQDKNSQQPCSRCSDGVIKLTPISAFSQDNIYEVQPPRVDRKSTEIFQAHIQASQGIMQPLGKEDSSMYREYIRNRYLEspecificaciones
NP_055660.1 | |
Liquid | |
9896 | |
FIG4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPTAAAPIISSVQKLVLYETRARYFLVGSNNAETKYRVLKIDRTEPKDLVIIDDRHVYTQQEVRELLGRLDLGNRTKMGQKGSSGLFRAVSAFGVVGFVRFLEGYYIVLITKRRKMADIGGHAIYKVEDTNMIYIPNDSVRVTHPDEARYLRIFQNVDLSSNFYFSYSYDLSHSLQYNLTVLRMPLEMLKSEMTQNRQESFDIFEDEGLITQGGSGVFGICSEPYMKYVWNGELLDIIKSTVHRDWLLYIIHGFCGQSKLLIYGRPVYVTLIARRSSKFAGTRFLKRGANCEGDVANEVETEQILCDASVMSFTAGSYSSYVQVRGSVPLYWSQDISTMMPKPPITLDQADPFAHVAALHFDQMFQRFGSPIIILNLVKEREKRKHERILSEELVAAVTYLNQFLPPEHTIVYIPWDMAKYTKSKLCNVLDRLNVIAESVVKKTGFFVNRPDSYCSILRPDEKWNELGGCVIPTGRLQTGILRTNCVDCLDRTNTAQFMVGKCALAYQLYSLGLIDKPNLQFDTDAVRLFEELYEDHGDTLSLQYGGSQLVHRVKTYRKIAPWTQHSKDIMQTLSRYYSNAFSDADRQDSINLFLGVFHPTEGKPHLWELPTDFYLHHKNTMRLLPTRRSYTYWWTPEVIKHLPLPYDEVICAVNLKKLIVKKFHKYEEEIDIHNEFFRPYELSSFDDTFCLAMTSSARDFMPKTVGIDPSPFTVRKPDETGKSVLGNKSNREEAVLQRKTAASAPPPPSEEAVSSSSEDDSGTDREEEGSVSQRSTPVKMTDAGDSAKVTENVVQPMKELYGINLSDGLSEEDFSIYSRFVQLGQSQHKQDKNSQQPCSRCSDGVIKLTPISAFSQDNIYEVQPPRVDRKSTEIFQAHIQASQGIMQPLGKEDSSMYREYIRNRYL | |
RUO | |
FIG4 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
130kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0274/SAC3/dJ249I4.1/hSac3 | |
FIG4 | |
Yes | |
wheat germ expression system |