missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FGF9 Partial ORF (NP_002001, 99 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_002001
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 2254
Peso molecular 37.84kDa
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16104951
Ver documentos Detalles de la promoción
Abnova™
H00002254-Q01.25UG
25 ug
508.00€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16194941
Ver documentos Detalles de la promoción
Abnova™
H00002254-Q01.10UG
10 ug
335.00€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis. [provided by RefSeq]

Sequence: SIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQS
Especificaciones

Especificaciones

NP_002001
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAF/HBFG-9/MGC119914/MGC119915
FGF9
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
2254
FGF9 (Human) Recombinant Protein (Q01)
SIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQS
RUO
FGF9
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Product Certifications