Learn More
Abnova™ Human FEZ1 Full-length ORF (NP_005094.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009638-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq]
Sequence: MEAPLVSLDEEFEDLRPSCSEDPEEKPQCFYGSSPHHLEDPSLSELENFSSEIISFKSMEDLVNEFDEKLNVCFRNYNAKTENLAPVKNQLQIQEEEETLQDEEVWDALTDNYIPSLSEDWRDPNIEALNGNCSDTEIHEKEEEEFNEKSENDSGINEEPLLTADQVIEEIEEMMQNSPDPEEEEEVLEEEDGGETSSQADSVLLQEMQALTQTFNNNWSYEGLRHMSGSELTELLDQVEGAIRDFSEELVQQLARRDELEFEKEVKNSFITVLIEVQNKQKEQRELMKKRRKEKGLSLQSSRIEKGNQMPLKRFSMEGISNILQSGIRQTFGSSGTDKQYLNTVIPYEKKASPPSVEDLQMLTNILFAMKEDNEKVPTLLTDYILKVLCPTEspecificaciones
NP_005094.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FEZ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9638 | |
FEZ1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEAPLVSLDEEFEDLRPSCSEDPEEKPQCFYGSSPHHLEDPSLSELENFSSEIISFKSMEDLVNEFDEKLNVCFRNYNAKTENLAPVKNQLQIQEEEETLQDEEVWDALTDNYIPSLSEDWRDPNIEALNGNCSDTEIHEKEEEEFNEKSENDSGINEEPLLTADQVIEEIEEMMQNSPDPEEEEEVLEEEDGGETSSQADSVLLQEMQALTQTFNNNWSYEGLRHMSGSELTELLDQVEGAIRDFSEELVQQLARRDELEFEKEVKNSFITVLIEVQNKQKEQRELMKKRRKEKGLSLQSSRIEKGNQMPLKRFSMEGISNILQSGIRQTFGSSGTDKQYLNTVIPYEKKASPPSVEDLQMLTNILFAMKEDNEKVPTLLTDYILKVLCPT | |
RUO | |
FEZ1 | |
Recombinant | |
wheat germ expression system |