Learn More
Abnova™ Human FBXW4 Partial ORF (NP_071322, 41 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006468-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the F-box/WD-40 gene family, which recruit specific target proteins through their WD-40 protein-protein binding domains for ubiquitin mediated degradation. In mouse, a highly similar protein is thought to be responsible for maintaining the apical ectodermal ridge of developing limb buds; disruption of the mouse gene results in the absence of central digits, underdeveloped or absent metacarpal/metatarsal bones and syndactyly. This phenotype is remarkably similar to split hand-split foot malformation in humans, a clinically heterogeneous condition with a variety of modes of transmission. An autosomal recessive form has been mapped to the chromosomal region where this gene is located, and complex rearrangements involving duplications of this gene and others have been associated with the condition. A pseudogene of this locus has been mapped to one of the introns of the BCR gene on chromosome 22. [provided by RefSeq]
Sequence: SYLDMRALGRLAQVCRWLRRFTSCDLLWRRIARASLNSGFTRLGTDLMTSVPVKERVKVSQNWRLGRCREGILLKWRCSQMPWMQLEDDSLYISQANFILEspecificaciones
NP_071322 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SYLDMRALGRLAQVCRWLRRFTSCDLLWRRIARASLNSGFTRLGTDLMTSVPVKERVKVSQNWRLGRCREGILLKWRCSQMPWMQLEDDSLYISQANFIL | |
RUO | |
FBXW4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6468 | |
FBXW4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAC/FBW4/FBWD4/SHFM3/SHSF3 | |
FBXW4 | |
Recombinant | |
wheat germ expression system |