missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO8 Full-length ORF (AAH14679, 1 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00026269-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198252

  • 302.00€ / 10 microgramos


Fecha estimada de envío
22-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MGQGLWRVVRNQQLQQEGYSEQGYLTREQSRRMAASNISNTNHRKQVQGGIDIYHLLKARKSKEQEGFINLEVLPPELSFTILSYLNATDLCLASCVWQDLANDELLWQGLCKSTWGHCSIYNKNPPLGFSFRKLYMQLDEGSLTFNANPDEGVNYFMSKGILDDSPKEIAKFIFCTRTLNWKKLRIYLDERRDVLDDLVTLHNFRNQFLPNALREFFRHIHAPEERGEYLETLITKFSHRFCACNPDLMRELGLSPDAVYVLCYSLILLSIDLTSPHVKNKMSKREFIRNTRRAAQNISEDFVGHLYDNIYLIGHVAA
Especificaciones

Especificaciones

AAH14679
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.83kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DC10|FBS|FBX8
FBXO8
Yes
wheat germ expression system
Liquid
26269
FBXO8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGQGLWRVVRNQQLQQEGYSEQGYLTREQSRRMAASNISNTNHRKQVQGGIDIYHLLKARKSKEQEGFINLEVLPPELSFTILSYLNATDLCLASCVWQDLANDELLWQGLCKSTWGHCSIYNKNPPLGFSFRKLYMQLDEGSLTFNANPDEGVNYFMSKGILDDSPKEIAKFIFCTRTLNWKKLRIYLDERRDVLDDLVTLHNFRNQFLPNALREFFRHIHAPEERGEYLETLITKFSHRFCACNPDLMRELGLSPDAVYVLCYSLILLSIDLTSPHVKNKMSKREFIRNTRRAAQNISEDFVGHLYDNIYLIGHVAA
RUO
FBXO8
Wheat Germ (in vitro)
GST
Videos
Documentos