missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXL21 Partial ORF (NP_036291, 167 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00026223-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108182

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains 6 tandem leucine-rich repeats. The amino acid sequence of this protein is highly similar to that of f-box and leucine-rich repeat protein 3A. Comparisons of this gene to orthologous sequences suggest that it may be a pseudogene, and may no longer express a functional protein.[provided by RefSeq]

Sequence: VSKVVLGRVGLNCPRLIELVVCANDLQPLDNELICIAEHCTNLTALGLSKCEVSCSAFIRFVRLCERRLTQLSVMEEVLIPDEDYSLDEIHTEVSKYLGRVWFPDVMPLW
Especificaciones

Especificaciones

NP_036291
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VSKVVLGRVGLNCPRLIELVVCANDLQPLDNELICIAEHCTNLTALGLSKCEVSCSAFIRFVRLCERRLTQLSVMEEVLIPDEDYSLDEIHTEVSKYLGRVWFPDVMPLW
RUO
FBXL21
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26223
FBXL21 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBL3B/FBXL3B/FBXL3P/Fbl21/MGC120237
FBXL21
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales