Learn More
Abnova™ Human FBXL18 Partial ORF (NP_079239, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080028-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the F-box protein family, such as FBXL18, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MASSGEDISNDDDDMHPAAAGMADGVHLLGFSDEILLHILSHVPSTDLILNVRRTCRKLAALCLDKSLIHTVLLQKDYQASEDKVRQLVKEIGREIQQLSMAGCYWLPGSEspecificaciones
NP_079239 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASSGEDISNDDDDMHPAAAGMADGVHLLGFSDEILLHILSHVPSTDLILNVRRTCRKLAALCLDKSLIHTVLLQKDYQASEDKVRQLVKEIGREIQQLSMAGCYWLPGS | |
RUO | |
FBXL18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80028 | |
FBXL18 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10776/FLJ11467/FLJ26934/FLJ38075/FLJ41541/Fbl18 | |
FBXL18 | |
Recombinant | |
wheat germ expression system |