missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FAM46D Full-length ORF (NP_689843.1, 1 a.a. - 389 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00169966-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16109113

  • 341.70€ / 10 microgramos
Envío estimado: 17-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sequence: MSEIRFTNLTWDQVITLDQVLDEVIPIHGKGNFPTMEVKPKDIIHVVKDQLIGQGIIVKDARLNGSVASYILASHNGISYKDLDVIFGVELPGNEEFQVVKDAVLDCLLDFLPKDVKKEKLSPDIMKDAYVQKLVKVCNGHDCWSLISLSNNTGKNLELKFVSSLRRQFEFSVDSFQIVLDPMLDFYSDKNAKLTKESYPVVVAESMYGDFQEAMTHLQHKLICTRKPEEIRGGGLLKYCSLLVHGFKPACMSEIKNLERYMCSRFFIDFPHIEEQQKKIESYLHNHFIGEGMTKYDYLMTLHGVVNESTVCLMSYERRQILHLITMMALKVLGELNILPNTQKVTCFYQPAPYFAAEARYPIYVIPEPPPVSFQPYHPLHFRGSNGMS
Especificaciones

Especificaciones

NP_689843.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC26999
FAM46D
Yes
wheat germ expression system
Liquid
169966
FAM46D (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSEIRFTNLTWDQVITLDQVLDEVIPIHGKGNFPTMEVKPKDIIHVVKDQLIGQGIIVKDARLNGSVASYILASHNGISYKDLDVIFGVELPGNEEFQVVKDAVLDCLLDFLPKDVKKEKLSPDIMKDAYVQKLVKVCNGHDCWSLISLSNNTGKNLELKFVSSLRRQFEFSVDSFQIVLDPMLDFYSDKNAKLTKESYPVVVAESMYGDFQEAMTHLQHKLICTRKPEEIRGGGLLKYCSLLVHGFKPACMSEIKNLERYMCSRFFIDFPHIEEQQKKIESYLHNHFIGEGMTKYDYLMTLHGVVNESTVCLMSYERRQILHLITMMALKVLGELNILPNTQKVTCFYQPAPYFAAEARYPIYVIPEPPPVSFQPYHPLHFRGSNGMS
RUO
FAM46D
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales