missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EYS Partial ORF (XP_498111, 309 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00346007-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16172857

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The product of this gene contains multiple epidermal growth factor (EGF)-like and LamG domains. The protein is expressed in the photoreceptor layer of the retina, and the gene is mutated in autosomal recessive retinitis pigmentosa. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: HVVVIQNQTLIKAYINNSLILSEDIDPHKNFVALNYDGICYLGGFEYGRKVNIVTQEIFKTNFVGKIKDVVFFQEPKNIELIKLEGYNVYDGDEQNEVT
Especificaciones

Especificaciones

XP_498111
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HVVVIQNQTLIKAYINNSLILSEDIDPHKNFVALNYDGICYLGGFEYGRKVNIVTQEIFKTNFVGKIKDVVFFQEPKNIELIKLEGYNVYDGDEQNEVT
RUO
EYS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
346007
EYS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf178/C6orf179/C6orf180/EGFL10/EGFL11/KIAA0663/RP25/SPAM/bA166P24.2/bA307F22.3/bA74E24.1/dJ1018A4.2/dJ22I17.2/dJ303F19.1
EYS
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales