missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EPN3 Partial ORF (NP_060427, 326 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055040-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: PGFRPNTEASGSSWGPSADPWSPIPSGTVLSRSQPWDLTPMLSSSEPWGRTPVLPAGPPTTDPWALNSPHYKLPSTGADPWGASLETSDTPGGASTFDPFAEspecificaciones
NP_060427 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PGFRPNTEASGSSWGPSADPWSPIPSGTVLSRSQPWDLTPMLSSSEPWGRTPVLPAGPPTTDPWALNSPHYKLPSTGADPWGASLETSDTPGGASTFDPFA | |
RUO | |
EPN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55040 | |
EPN3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20778/MGC129899 | |
EPN3 | |
Recombinant | |
wheat germ expression system |