Learn More
ACROBiosystems Human Ephrin-A4 / EFNA4 Protein, Fc Tag
Marca: ACROBiosystems EA4-H5250-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Ephrin-A4 (EFNA4) is also known as EPH-related receptor tyrosine kinase ligand 4 (LERK4), EPLG4, which belongs to the ephrin family. EFNA4 contains one ephrin RBD (ephrin receptor-binding) domain. EFNA4 is cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. EFNA4 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. EFNA4 may play a role in the interaction between activated B-lymphocytes and dendritic cells in tonsils.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.0 kDa. The protein migrates as 45-50 kDa and 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAI07484 | |
43.0 kDa | |
1 mg | |
Leu 26 - Gly 171 | |
EFNA4,EPLG4,LERK4,Ephrin-A4 | |
Unconjugated | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484). | |
-20°C | |
Ephrin-A4 | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |