missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EML1 Partial ORF (NP_001008707, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002009-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16107634

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Human echinoderm microtubule-associated protein-like is a strong candidate for the Usher syndrome type 1A gene. Usher syndromes (USHs) are a group of genetic disorders consisting of congenital deafness, retinitis pigmentosa, and vestibular dysfunction of variable onset and severity depending on the genetic type. The disease process in USHs involves the entire brain and is not limited to the posterior fossa or auditory and visual systems. The USHs are catagorized as type I (USH1A, USH1B, USH1C, USH1D, USH1E and USH1F), type II (USH2A and USH2B) and type III (USH3). The type I is the most severe form. Gene loci responsible for these three types are all mapped. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MEDGFSSYSSLYDTSSLLQFCNDDSASAASSMEVTDRIASLEQRVQMQEDDIQLLKSALADVVRRLNITEEQQAVLNRKGPTKARPLMQTLPLRTTVNN
Especificaciones

Especificaciones

NP_001008707
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEDGFSSYSSLYDTSSLLQFCNDDSASAASSMEVTDRIASLEQRVQMQEDDIQLLKSALADVVRRLNITEEQQAVLNRKGPTKARPLMQTLPLRTTVNN
RUO
EML1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2009
EML1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELP79/EMAP/EMAPL/FLJ45033/HuEMAP
EML1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales