missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUSP15 Full-length ORF (AAH56911.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00128853-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16197823

  • 295.00€ / 10 microgramos


Fecha estimada de envío
23-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MGNGMTKVLPGLYLGNFIDAKDLDQLGRNKITHIISIHESPQPLLQDITYLRIPVADTPEVPIKKHFKECINFIHCCRLNGGNCLVHCFAGISRSTTIVTAYVMTVTGLGWRDVLEAIKATRPIANPNPGFRQQLEEFGWASSQKLRRQLEERFGESPFRDEEELRALLPLCKRCRQGSATSASSAGPHSAASEGTLQRLVPRTPREAHRPLPLLARVKQTFSCLPRCLSRKGGK
Especificaciones

Especificaciones

AAH56911.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ40111|VHY
DUSP15
Yes
wheat germ expression system
Liquid
128853
DUSP15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNGMTKVLPGLYLGNFIDAKDLDQLGRNKITHIISIHESPQPLLQDITYLRIPVADTPEVPIKKHFKECINFIHCCRLNGGNCLVHCFAGISRSTTIVTAYVMTVTGLGWRDVLEAIKATRPIANPNPGFRQQLEEFGWASSQKLRRQLEERFGESPFRDEEELRALLPLCKRCRQGSATSASSAGPHSAASEGTLQRLVPRTPREAHRPLPLLARVKQTFSCLPRCLSRKGGK
RUO
DUSP15
Wheat Germ (in vitro)
GST
Videos
Documentos