Learn More
Abnova™ Human DSTN Full-length ORF (AAH09477.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011034-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MASGVQVADEVCRIFYDMKVRKCSTPEEIKKRKKAVIFCLSADKKCIIVEEGKEILVGDVGVTITDPFKHFVGMLPEKDCRYALYDASFETKESRKEELMFFLWAPELAPLKSKMIYASSKDAIKKKFQGIKHECQANGPEDLNRACIAEKLGGSLIVAFEGCPVEspecificaciones
AAH09477.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACTDP/ADF/bA462D18.2 | |
DSTN | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11034 | |
DSTN (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASGVQVADEVCRIFYDMKVRKCSTPEEIKKRKKAVIFCLSADKKCIIVEEGKEILVGDVGVTITDPFKHFVGMLPEKDCRYALYDASFETKESRKEELMFFLWAPELAPLKSKMIYASSKDAIKKKFQGIKHECQANGPEDLNRACIAEKLGGSLIVAFEGCPV | |
RUO | |
DSTN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |