Learn More
Abnova™ Human DRIM Partial ORF (NP_055318, 946 a.a. - 1053 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027340-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
UTP20 is a component of the U3 small nucleolar RNA (snoRNA) (SNORD3A; MIM 180710) protein complex (U3 snoRNP) and is involved in 18S rRNA processing (Wang et al., 2007 [PubMed 17498821]).[supplied by OMIM]
Sequence: LHQDQMVQKITLDCIMTYKHPHVLPYRENLQRLLEDRSFKEEIVHFSISEDNAVVKTAHRADLFPILMRILYGRMKNKTGSKTQGKSASGTRMAIVLRFLAGTQPEEIEspecificaciones
NP_055318 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LHQDQMVQKITLDCIMTYKHPHVLPYRENLQRLLEDRSFKEEIVHFSISEDNAVVKTAHRADLFPILMRILYGRMKNKTGSKTQGKSASGTRMAIVLRFLAGTQPEEI | |
RUO | |
UTP20 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27340 | |
DRIM (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRIM | |
UTP20 | |
Recombinant | |
wheat germ expression system |