Learn More
ACROBiosystems Human DR3 / TNFRSF25 Protein, His Tag
Marca:  ACROBiosystems TN5-H52H3-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17206590
  • 2486.22€ / 1 miligramo
Fecha estimada de envío: 20-10-2022
para ver el stock
Descripción
Descripción
Tumor necrosis factor receptor superfamily member 25 (TNFRSF25) is also known as Apo-3, Death receptor 3 (DDR3 or DR3), Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor TRAMP, Lymphocyte-associated receptor of death, Apo-3, which is a member of the TNF-receptor superfamily. TNFRSF25 is a homodimer protein, which can Interact strongly via the death domains with TNFRSF1 and TRADD to activate at least two distinct signaling cascades, apoptosis and NF-kappa-B signaling. TNFRSF25 is receptor for TNFSF12 / APO3L / TWEAK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.8 kDa.
Especificaciones
Especificaciones
Powder
1 mg
Gln 25 - Gln 199
TNFRSF25,DR3,APO3,DDR3,TNFRSF12,WSL,WSL1
Unconjugated
HEK293
Human
20.8 kDa
Human DR3 / TNFRSF25, His Tag (TN5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 199 (Accession # Q93038-1).
-20°C
DR3 / TNFRSF25
His Tag
Other Recombinant Proteins