missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOCK7 Full-length ORF (AAH16392.1, 1 a.a. - 624 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00085440-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166333

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 04-05-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sequence: MACNQSAVYLQHCFATQRALVSKFPELLFEEETEQCADLCLRLLRHCSSSIGTIRSHASASLYLLMRQNFEIGNNFARVKMQVTMSLSSLVGTSQNFNEEFLRRSLKTILTYAEEDLELRETTFPDQVQDLVFNLHMILSDTVKMKEHQEDPEMLIDLMYRIAKGYQTSPDLRLTWLQNMAGKHSERSNHAEAAQCLVHSAALVAEYLSMLEDRKYLPVGCVTFQNISSNVLEESAVSDDVVSPDEEGICSGKYFTESGLVGLLEQAAASFSMAGMYEAVNEVYKVLIPIHEANRDAKKLSTIHGKLQEAFSKIVHQDGKRMFGTYFRVGFYGTKFGDLDEQEFVYKEPAITKLAEISHRLEGFYGERFGEDVVEVIKDSNPVDKCKLDPNKAYIQITYVEPYFDTYEMKDRITYFDKNYNLRRFMYCTPFTLDGRAHGELHEQFKRKTILTTSHAFPYIKTRVNVTHKEEIILTPIEVAIEDMQKKTQELAFATHQDPADPKMLQMVLQGSVGTTVNQGPLEVAQVFLSEIPSDPKLFRHHNKLRLCFKDFTKRCEDALRKNKSLIGPDQKEYQRELERNYHRLKEALQPLINRKIPQLYKAVLPVTCHRDSFSRMSLRKMDL
Especificaciones

Especificaciones

AAH16392.1
Liquid
85440
DOCK7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MACNQSAVYLQHCFATQRALVSKFPELLFEEETEQCADLCLRLLRHCSSSIGTIRSHASASLYLLMRQNFEIGNNFARVKMQVTMSLSSLVGTSQNFNEEFLRRSLKTILTYAEEDLELRETTFPDQVQDLVFNLHMILSDTVKMKEHQEDPEMLIDLMYRIAKGYQTSPDLRLTWLQNMAGKHSERSNHAEAAQCLVHSAALVAEYLSMLEDRKYLPVGCVTFQNISSNVLEESAVSDDVVSPDEEGICSGKYFTESGLVGLLEQAAASFSMAGMYEAVNEVYKVLIPIHEANRDAKKLSTIHGKLQEAFSKIVHQDGKRMFGTYFRVGFYGTKFGDLDEQEFVYKEPAITKLAEISHRLEGFYGERFGEDVVEVIKDSNPVDKCKLDPNKAYIQITYVEPYFDTYEMKDRITYFDKNYNLRRFMYCTPFTLDGRAHGELHEQFKRKTILTTSHAFPYIKTRVNVTHKEEIILTPIEVAIEDMQKKTQELAFATHQDPADPKMLQMVLQGSVGTTVNQGPLEVAQVFLSEIPSDPKLFRHHNKLRLCFKDFTKRCEDALRKNKSLIGPDQKEYQRELERNYHRLKEALQPLINRKIPQLYKAVLPVTCHRDSFSRMSLRKMDL
RUO
DOCK7
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
98.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1771/ZIR2
DOCK7
Yes
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales