Learn More
Abnova™ Human DNAJC6 Full-length ORF (NP_055602.1, 1 a.a. - 913 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009829-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
DNAJC6 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus, a glycine/phenylalanine (G/F)-rich region, and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]
Sequence: MKDSENKGASSPDMEPSYGGGLFDMVKGGAGRLFSNLKDNLKDTLKDTSSRVIQSVTSYTKGDLDFTYVTSRIIVMSFPLDNVDIGFRNQVDDIRSFLDSRHLDHYTVYNLSPKSYRTAKFHSRVSECSWPIRQAPSLHNLFAVCRNMYNWLLQNPKNVCVVHCLDGRAASSILVGAMFIFCNLYSTPGPAIRLLYAKRPGIGLSPSHRRYLGYMCDLLADKPYRPHFKPLTIKSITVSPIPFFNKQRNGCRPYCDVLIGETKIYSTCTDFERMKEYRVQDGKIFIPLNITVQGDVVVSMYHLRSTIGSRLQAKVTNTQIFQLQFHTGFIPLDTTVLKFTKPELDACDVPEKYPQLFQVTLDVELQPHDKVIDLTPPWEHYCTKDVNPSILFSSHQEHQDTLALGGQAPIDIPPDNPRHYGQSGFFASLCWQDQKSEKSFCEEDHAALVNQESEQSDDELLTLSSPHGNANGDKPHGVKKPSKKQQEPAAPPPPEDVDLLGLEGSAMSNSFSPPAAPPTNSELLSDLFGGGGAAGPTQAGQSGVEDVFHPSGPASTQSTPRRSATSTSASPTLRVGEGATFDPFGAPSKPSGQDLLGSFLNTSSASSDPFLQPTRSPSPTVHASSTPAVNIQPDVSGGWDWHAKPGGFGMGSKSAATSPTGSSHGTPTHQSKPQTLDPFADLGTLGSSSFASKPTTPTGLGGGFPPLSSPQKASPQPMGGGWQQGGAYNWQQPQPKPQPSMPHSSPQNRPNYNVSFSAMPGGQNERGKGSSNLEGKQKAADFEDLLSGQGFNAHKDKKGPRTIAEMRKEEMAKEMDPEKLKILEWIEGKERNIRALLSTMHTVLWAGETKWKPVGMADLVTPEQVKKVYRKAVLVVHPDKATGQPYEQYAKMIFMELNDAWSEFENQGQKPLYEspecificaciones
NP_055602.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
126.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJC6/KIAA0473/MGC129914/MGC129915/MGC48436 | |
DNAJC6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9829 | |
DNAJC6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKDSENKGASSPDMEPSYGGGLFDMVKGGAGRLFSNLKDNLKDTLKDTSSRVIQSVTSYTKGDLDFTYVTSRIIVMSFPLDNVDIGFRNQVDDIRSFLDSRHLDHYTVYNLSPKSYRTAKFHSRVSECSWPIRQAPSLHNLFAVCRNMYNWLLQNPKNVCVVHCLDGRAASSILVGAMFIFCNLYSTPGPAIRLLYAKRPGIGLSPSHRRYLGYMCDLLADKPYRPHFKPLTIKSITVSPIPFFNKQRNGCRPYCDVLIGETKIYSTCTDFERMKEYRVQDGKIFIPLNITVQGDVVVSMYHLRSTIGSRLQAKVTNTQIFQLQFHTGFIPLDTTVLKFTKPELDACDVPEKYPQLFQVTLDVELQPHDKVIDLTPPWEHYCTKDVNPSILFSSHQEHQDTLALGGQAPIDIPPDNPRHYGQSGFFASLCWQDQKSEKSFCEEDHAALVNQESEQSDDELLTLSSPHGNANGDKPHGVKKPSKKQQEPAAPPPPEDVDLLGLEGSAMSNSFSPPAAPPTNSELLSDLFGGGGAAGPTQAGQSGVEDVFHPSGPASTQSTPRRSATSTSASPTLRVGEGATFDPFGAPSKPSGQDLLGSFLNTSSASSDPFLQPTRSPSPTVHASSTPAVNIQPDVSGGWDWHAKPGGFGMGSKSAATSPTGSSHGTPTHQSKPQTLDPFADLGTLGSSSFASKPTTPTGLGGGFPPLSSPQKASPQPMGGGWQQGGAYNWQQPQPKPQPSMPHSSPQNRPNYNVSFSAMPGGQNERGKGSSNLEGKQKAADFEDLLSGQGFNAHKDKKGPRTIAEMRKEEMAKEMDPEKLKILEWIEGKERNIRALLSTMHTVLWAGETKWKPVGMADLVTPEQVKKVYRKAVLVVHPDKATGQPYEQYAKMIFMELNDAWSEFENQGQKPLY | |
RUO | |
DNAJC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |