missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DNAJB7 Full-length ORF (NP_660157.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00150353-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108613

  • 518.16€ / 25 microgramos
Envío estimado: 24-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

DNAJB7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]

Sequence: MVDYYEVLGLQRYASPEDIKKAYHKVALKWHPDKNPENKEEAERKFKEVAEAYEVLSNDEKRDIYDKYGTEGLNGGGSHFDDECEYGFTFHKPDDVFKEIFHERDPFSFHFFEDSLEDLLNRPGSSYGNRNRDAGYFFSTASEYPIFEKFSSYDTGYTSQGSLGHEGLTSFSSLAFDNSGMDNYISVTTSDKIVNGRNINTKKIIESDQEREAEDNGELTFFLVNSVANEEGFAKECSWRTQSFNNYSPNSHSSKHVSQYTFVDNDEGGISWVTSNRDPPIFSAGVKEGGKRKKKKRKEVQKKSTKRNC
Especificaciones

Especificaciones

NP_660157.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ5/HSC3/MGC138340
DNAJB7
Yes
wheat germ expression system
Liquid
150353
DNAJB7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVDYYEVLGLQRYASPEDIKKAYHKVALKWHPDKNPENKEEAERKFKEVAEAYEVLSNDEKRDIYDKYGTEGLNGGGSHFDDECEYGFTFHKPDDVFKEIFHERDPFSFHFFEDSLEDLLNRPGSSYGNRNRDAGYFFSTASEYPIFEKFSSYDTGYTSQGSLGHEGLTSFSSLAFDNSGMDNYISVTTSDKIVNGRNINTKKIIESDQEREAEDNGELTFFLVNSVANEEGFAKECSWRTQSFNNYSPNSHSSKHVSQYTFVDNDEGGISWVTSNRDPPIFSAGVKEGGKRKKKKRKEVQKKSTKRNC
RUO
DNAJB7
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales