Learn More
Abnova™ Human DNAJB12 Partial ORF (NP_060096, 10 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054788-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]
Sequence: RCISIALKAIQSNQPDRALRFLEKAQRLYPTPRVRALIESLNQKPQTAGDQPPPTDTTHATHRKAGGTDAPSANGEAGGESTKGYTAEQVAAVKRVKQCKDYYEILGVSREspecificaciones
NP_060096 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RCISIALKAIQSNQPDRALRFLEKAQRLYPTPRVRALIESLNQKPQTAGDQPPPTDTTHATHRKAGGTDAPSANGEAGGESTKGYTAEQVAAVKRVKQCKDYYEILGVSR | |
RUO | |
DNAJB12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54788 | |
DNAJB12 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ10/DKFZp586B2023 | |
DNAJB12 | |
Recombinant | |
wheat germ expression system |