missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DIMT1L Full-length ORF (NP_055288.1, 1 a.a. - 313 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027292-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MPKVKSGAIGRRRGRQEQRRELKSAGGLMFNTGIGQHILKNPLIINSIIDKAALRPTDVVLEVGPGTGNMTVKLLEKAKKVVACELDPRLVAELHKRVQGTPVASKLQVLVGDVLKTDLPFFDTCVANLPYQISSPFVFKLLLHRPFFRCAILMFQREFALRLVAKPGDKLYCRLSINTQLLARVDHLMKVGKNNFRPPPKVESSVVRIEPKNPPPPINFQEWDGLVRITFVRKNKTLSAAFKSSAVQQLLEKNYRIHCSVHNIIIPEDFSIADKIQQILTSTGFSDKRARSMDIDDFIRLLHGFNAEGIHFSEspecificaciones
NP_055288.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DIM1/DIMT1/HSA9761/HUSSY5 | |
DIMT1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27292 | |
DIMT1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPKVKSGAIGRRRGRQEQRRELKSAGGLMFNTGIGQHILKNPLIINSIIDKAALRPTDVVLEVGPGTGNMTVKLLEKAKKVVACELDPRLVAELHKRVQGTPVASKLQVLVGDVLKTDLPFFDTCVANLPYQISSPFVFKLLLHRPFFRCAILMFQREFALRLVAKPGDKLYCRLSINTQLLARVDHLMKVGKNNFRPPPKVESSVVRIEPKNPPPPINFQEWDGLVRITFVRKNKTLSAAFKSSAVQQLLEKNYRIHCSVHNIIIPEDFSIADKIQQILTSTGFSDKRARSMDIDDFIRLLHGFNAEGIHFS | |
RUO | |
DIMT1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |