missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGAT2 Full-length ORF (NP_115953.2, 1 a.a. - 388 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

en la oferta

Especificaciones

Nombre DGAT2 (Human) Recombinant Protein (P01)
Recombinante Yes
Método de purificación Glutathione Sepharose 4 Fast Flow
Peso molecular 70.2kDa
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ver más especificaciones

Descripción

Descripción

Acyl-CoA:diacylglycerol acyltransferase, or DGAT (EC 2.3.1.20), is responsible for the synthesis of triglycerides. It catalyzes a reaction in which diacylglycerol is covalently joined to long chain fatty acyl-CoAs.[supplied by OMIM]

Sequence: MKTLIAAYSGVLRGERQAEADRSQRSHGGPALSREGSGRWGTGSSILSALQDLFSVTWLNRSKVEKQLQVISVLQWVLSFLVLGVACSAILMYIFCTDCWLIAVLYFTWLVFDWNTPKKGGRRSQWVRNWAVWRYFRDYFPIQLVKTHNLLTTRNYIFGYHPHGIMGLGAFCNFSTEATEVSKKFPGIRPYLATLAGNFRMPVLREYLMSGGICPVSRDTIDYLLSKNGSGNAIIIVVGGAAESLSSMPGKNAVTLRNRKGFVKLALRHGADLVPIYSFGENEVYKQVIFEEGSWGRWVQKKFQKYIGFAPCIFHGRGLFSSDTWGLVPYSKPITTVVGEPITIPKLEHPTQQDIDLYHTMYMEALVKLFDKHKTKFGLPETEVLEVN
Especificaciones

Especificaciones

DGAT2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
70.2kDa
Videos
SDS
Documentos

Documentos

Product Certifications