missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Dermatopontin/TRAMP ELISA Kit

Human Dermatopontin/TRAMP quantitates human Dermatopontin/TRAMP in serum, plasma, supernatant.

Marca:  Invitrogen™ EH151RBX10

Detalles adicionales : Peso : 2.50000kg

 Ver más versiones de este producto

Código de producto. 17804080

  • 4540.00€ / paquete de 10

Fecha estimada de envío: 05-05-2023
para ver el stock
Añadir a la cesta

Descripción

Descripción

Human Dermatopontin/TRAMP quantitates human Dermatopontin/TRAMP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Dermatopontin/TRAMP.

Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.
Especificaciones

Especificaciones

1810032B19Rik; 5033416F05Rik; Dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; TRAMP; Tyrosine-rich acidic matrix protein
0.43 ng/mL
ELISA
Colorimetric Microplate Reader
DPT
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.43 to 100 ng/mL
Biotin
Supernatant
Q07507
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
DPT
4 hr 45 min
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales