missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DDX59 Full-length ORF (NP_001026895.1, 1 a.a. - 619 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00083479-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MFVPRSLKIKRNANDDGKSCVAKIIKPDPEDLQLDKSRDVPVDAVATEAATIDRHISESCPFPSPGGQLAEVHSVSPEQGAKDSHPSEEPVKSFSKTQRWAEPGEPICVVCGRYGEYICDKTDEDVCSLECKAKHLLQVKEKEEKSKLSNPQKADSEPESPLNASYVYKEHPFILNLQEDQIENLKQQLGILVQGQEVTRPIIDFEHCSLPEVLNHNLKKSGYEVPTPIQMQMIPVGLLGRDILASADTGSGKTAAFLLPVIMRALFESKTPSALILTPTRELAIQIERQAKELMSGLPRMKTVLLVGGLPLPPQLYRLQQHVKVIIATPGRLLDIIKQSSVELCGVKIVVVDEADTMLKMGFQQQVLDILENIPNDCQTILVSATIPTSIEQLASQLLHNPVRIITGEKNLPCANVRQIILWVEDPAKKKKLFEILNDKKLFKPPVLVFVDCKLGADLLSEAVQKITGLKRISIHSEKSQIERKNILKGLLEGDYEVVVSTGVLGRGLDLISVRLVVNFDMPSSMDEYVHQIGRVGRLGQNGTAITFINNNSKRLFWDIAKRVKPTGSILPPQLLNSPYLHDQKRKEQQKDKQTQNDLVTGANLMDIIRKHDKSNSQKEspecificaciones
NP_001026895.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564B1023/ZNHIT5 | |
DDX59 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83479 | |
DDX59 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFVPRSLKIKRNANDDGKSCVAKIIKPDPEDLQLDKSRDVPVDAVATEAATIDRHISESCPFPSPGGQLAEVHSVSPEQGAKDSHPSEEPVKSFSKTQRWAEPGEPICVVCGRYGEYICDKTDEDVCSLECKAKHLLQVKEKEEKSKLSNPQKADSEPESPLNASYVYKEHPFILNLQEDQIENLKQQLGILVQGQEVTRPIIDFEHCSLPEVLNHNLKKSGYEVPTPIQMQMIPVGLLGRDILASADTGSGKTAAFLLPVIMRALFESKTPSALILTPTRELAIQIERQAKELMSGLPRMKTVLLVGGLPLPPQLYRLQQHVKVIIATPGRLLDIIKQSSVELCGVKIVVVDEADTMLKMGFQQQVLDILENIPNDCQTILVSATIPTSIEQLASQLLHNPVRIITGEKNLPCANVRQIILWVEDPAKKKKLFEILNDKKLFKPPVLVFVDCKLGADLLSEAVQKITGLKRISIHSEKSQIERKNILKGLLEGDYEVVVSTGVLGRGLDLISVRLVVNFDMPSSMDEYVHQIGRVGRLGQNGTAITFINNNSKRLFWDIAKRVKPTGSILPPQLLNSPYLHDQKRKEQQKDKQTQNDLVTGANLMDIIRKHDKSNSQK | |
RUO | |
DDX59 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |