Learn More
Abnova™ Human DDX20 Full-length ORF (AAH34953, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011218-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which has an ATPase activity and is a component of the survival of motor neurons (SMN) complex. This protein interacts directly with SMN, the spinal muscular atrophy gene product, and may play a catalytic role in the function of the SMN complex on RNPs. [provided by RefSeq]
Sequence: MAAAFEASGALAAVATAMPAEHVAVQVPAPEPTPGPVRILRTAQDLSSPRTRTGDVLLAEPADFESLLLSRPVLEGLRAAGFERPSPVQLKAIPLGRCGLDLIVQAKSGTGKTCVFSTIALDSLVLENLSTQILILAPTREIAVQIHSVITAIGIKMEGLECHVFIGGTPLSQDKTRLKKCHIAVGSPGRIKQLIELDYLNPGSIRLFILDEADKLLEEGSFQEQINWIYSSLPASKQMLAVSATYPEFLANALTKYMRDPTFVRLNSSDPSLIGLKQYYKVVNSYPLAHKVFEEKTQHLQELFSRIPFNQALVFSNLHSRAQHLADILSSKGFPAECISGNMNQNQRLDAMAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEGNMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDHMASSRNNSVSGLSVKSKNNTKQKLPVKSHSECGIIEKATSPKELGCDRQSEEQMKNSVQTPVENSTNSQHQVKEALPVSLPQIPCLSSFKIHQPYTLTFAELVEDYEHYIKEGLEKPVEIIRHYTGPGDQTVNPQNGFVRNKVTEQRVPVLASSSQSGDSESDSDSYSSRTSSQSKGNKSYLEGSSDNQLKDSESTPVDDRISLEQPPNGSDTPNPEKYQESPGIQMKTRLKEGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQEspecificaciones
AAH34953 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
116.38kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434H052/DP103/GEMIN3 | |
DDX20 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11218 | |
DDX20 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAFEASGALAAVATAMPAEHVAVQVPAPEPTPGPVRILRTAQDLSSPRTRTGDVLLAEPADFESLLLSRPVLEGLRAAGFERPSPVQLKAIPLGRCGLDLIVQAKSGTGKTCVFSTIALDSLVLENLSTQILILAPTREIAVQIHSVITAIGIKMEGLECHVFIGGTPLSQDKTRLKKCHIAVGSPGRIKQLIELDYLNPGSIRLFILDEADKLLEEGSFQEQINWIYSSLPASKQMLAVSATYPEFLANALTKYMRDPTFVRLNSSDPSLIGLKQYYKVVNSYPLAHKVFEEKTQHLQELFSRIPFNQALVFSNLHSRAQHLADILSSKGFPAECISGNMNQNQRLDAMAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEGNMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDHMASSRNNSVSGLSVKSKNNTKQKLPVKSHSECGIIEKATSPKELGCDRQSEEQMKNSVQTPVENSTNSQHQVKEALPVSLPQIPCLSSFKIHQPYTLTFAELVEDYEHYIKEGLEKPVEIIRHYTGPGDQTVNPQNGFVRNKVTEQRVPVLASSSQSGDSESDSDSYSSRTSSQSKGNKSYLEGSSDNQLKDSESTPVDDRISLEQPPNGSDTPNPEKYQESPGIQMKTRLKEGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQ | |
RUO | |
DDX20 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |