missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CYB5R1 Partial ORF (NP_057327.2, 207 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051706-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: QCFLLFANQTEKDIILREDLEELQARYPNRFKLWFTLDHPPKDWAYSKGFVTADMIREHLPAPGDDVLVLLCGPPPMVQLACHPNLDKLGYSQKMRFTYEspecificaciones
NP_057327.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QCFLLFANQTEKDIILREDLEELQARYPNRFKLWFTLDHPPKDWAYSKGFVTADMIREHLPAPGDDVLVLLCGPPPMVQLACHPNLDKLGYSQKMRFTY | |
RUO | |
CYB5R1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51706 | |
CYB5R1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B5R.1/NQO3A2/humb5R2 | |
CYB5R1 | |
Recombinant | |
wheat germ expression system |