missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CTCFL Partial ORF (NP_542185, 193 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140690-Q01.25ug
Additional Details : Peso : 0.00010kg
Description
Sequence: AERTKEQLFFVETMSGDERSDEIVLTVSNSNVEEQEDQPTAGQADAEKAKSTKNQRKTKGAKGTFHCDVCMFTSSRMSSFNRHMKTHTSEKPHLCHLCLKTSpecifications
NP_542185 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AERTKEQLFFVETMSGDERSDEIVLTVSNSNVEEQEDQPTAGQADAEKAKSTKNQRKTKGAKGTFHCDVCMFTSSRMSSFNRHMKTHTSEKPHLCHLCLKT | |
RUO | |
CTCFL | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
140690 | |
CTCFL (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BORIS|CTCF-T|MGC163358|dJ579F20.2 | |
CTCFL | |
Yes |