missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CSNK1G3 Partial ORF (AAH47567.1, 307 a.a. - 423 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001456-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: LFTDLFDRKGYMFDYEYDWIGKQLPTPVGAVQQDPALSSNREAHQHRDKMQQSKNQVVSSTNGELNTDDPTAGRSNAPITAPTEVEVMDETNCQKVLNMWCCCFFKRRKRKTIQRHKEspecificaciones
AAH47567.1 | |
Liquid | |
1456 | |
CSNK1G3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSNK1G3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.50kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LFTDLFDRKGYMFDYEYDWIGKQLPTPVGAVQQDPALSSNREAHQHRDKMQQSKNQVVSSTNGELNTDDPTAGRSNAPITAPTEVEVMDETNCQKVLNMWCCCFFKRRKRKTIQRHK | |
RUO | |
CSNK1G3 | |
Yes | |
wheat germ expression system |