missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CREB5 Partial ORF (NP_001011666, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009586-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MFCTSGGNSASVMSMRPVPGSLSSLLHLHNRQRQPMPASMPGTLPNPTMPGSSAVLMPMERQMSVNSSIMGMQGPNLSNPCASPQVQPMHSEAKMRLKAEspecificaciones
NP_001011666 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFCTSGGNSASVMSMRPVPGSLSSLLHLHNRQRQPMPASMPGTLPNPTMPGSSAVLMPMERQMSVNSSIMGMQGPNLSNPCASPQVQPMHSEAKMRLKA | |
RUO | |
CREB5 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
9586 | |
CREB5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRE-BPA | |
CREB5 | |
Yes |