missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CREB3L4 Partial ORF (NP_570968.1, 324 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00148327-Q02.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: SEDYQPHGVTSRNILTHKDVTENLETQVVESRLREPPGAKDANGSTRTLLEKMGGKPRPSGRIRSVLHADEMEspecificaciones
NP_570968.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEDYQPHGVTSRNILTHKDVTENLETQVVESRLREPPGAKDANGSTRTLLEKMGGKPRPSGRIRSVLHADEM | |
RUO | |
CREB3L4 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
148327 | |
CREB3L4 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIBZIP|ATCE1|CREB3|CREB4|JAL|hJAL | |
CREB3L4 | |
Yes |