Learn More
Abnova™ Human CPOX Partial ORF (NP_000088, 356 a.a. - 453 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001371-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Coproporphyrinogen oxidase (EC 1.3.3.3) is the sixth enzyme of the heme biosynthetic pathway. This soluble protein is localized in the intermembrane space of mitochondria and catalyzes the stepwise oxidative decarboxylation of the heme precursor coproporphyrinogen III to protoporphyrinogen IX via the tricarboxylic intermediate harderoporphyrinogen.[supplied by OMIM]
Sequence: SCARAVVPSYIPLVKKHCDDSFTPQEKLWQQLRRGRYVEFNLLYDRGTKFGLFTPGSRIESILMSLPLTARWEYMHSPSENSKEAEILEVLRHPRDWVEspecificaciones
NP_000088 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SCARAVVPSYIPLVKKHCDDSFTPQEKLWQQLRRGRYVEFNLLYDRGTKFGLFTPGSRIESILMSLPLTARWEYMHSPSENSKEAEILEVLRHPRDWV | |
RUO | |
CPOX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1371 | |
CPOX (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPO/CPX/HCP | |
CPOX | |
Recombinant | |
wheat germ expression system |