missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CPNE7 Partial ORF (NP_705900, 460 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00027132-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16105736

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ASRLPMSIIIVGVGNADFTDMQVLDGDDGVLRSPRGEPALRDIVQFVPFRELKNASPAALAKCVLAEVPKQVVEYYSHRGLPPRSLGVPAGEASPGCTP
Especificaciones

Especificaciones

NP_705900
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASRLPMSIIIVGVGNADFTDMQVLDGDDGVLRSPRGEPALRDIVQFVPFRELKNASPAALAKCVLAEVPKQVVEYYSHRGLPPRSLGVPAGEASPGCTP
RUO
CPNE7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27132
CPNE7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34192
CPNE7
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales