missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CPA5 Full-length ORF (NP_525124.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00093979-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16106863

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Carboxypeptidases have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Members of the A/B subfamily of carboxypeptidases, such as CPA5, contain an approximately 90-amino acid pro region that assists in the folding of the active carboxypeptidase domain. Cleavage of the pro region activates the enzyme (Wei et al., 2002 [PubMed 11836249]).[supplied by OMIM]

Sequence: MQGTPGGGTRPGPSPVDRRTLLVFSFILAAALGQMNFTGDQVLRVLAKDEKQLSLLGDLEGLKPQKVDFWRGPARPSLPVDMRVPFSELKDIKAYLESHGLAYSIMIKDIQVLLDEERQAMAKSRRLERSTNSFSYSSYHTLEEIYSWIDNFVMEHSDIVSKIQIGNSFENQSILVLKFSTGGSRHPAIWIDTGIHSREWITHATGIWTANKIVSDYGKDRVLTDILNAMDIFIELVTNPDGFAFTHSMNRLWRKNKSIRPGIFCIGVDLNRNWKSGFGGNGSNSNPCSETYHGPSPQSEPEVAAIVNFITAHGNFKALISIHSYSQMLMYPYGRSLDPVSNQRELYDLAKDAVEALYKVHGIEYIFGSISTTLYVASGITVDWAYDSGIKYAFSFELRDTGQYGFLLPATQIIPTAQETWMALRTIMEHTLNHPY
Especificaciones

Especificaciones

NP_525124.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35969
CPA5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
93979
CPA5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQGTPGGGTRPGPSPVDRRTLLVFSFILAAALGQMNFTGDQVLRVLAKDEKQLSLLGDLEGLKPQKVDFWRGPARPSLPVDMRVPFSELKDIKAYLESHGLAYSIMIKDIQVLLDEERQAMAKSRRLERSTNSFSYSSYHTLEEIYSWIDNFVMEHSDIVSKIQIGNSFENQSILVLKFSTGGSRHPAIWIDTGIHSREWITHATGIWTANKIVSDYGKDRVLTDILNAMDIFIELVTNPDGFAFTHSMNRLWRKNKSIRPGIFCIGVDLNRNWKSGFGGNGSNSNPCSETYHGPSPQSEPEVAAIVNFITAHGNFKALISIHSYSQMLMYPYGRSLDPVSNQRELYDLAKDAVEALYKVHGIEYIFGSISTTLYVASGITVDWAYDSGIKYAFSFELRDTGQYGFLLPATQIIPTAQETWMALRTIMEHTLNHPY
RUO
CPA5
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales