missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COX6B2 Full-length ORF (NP_653214.2, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal
Liquid
Marca: Abnova™ H00125965-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MLDVEAQEPPKGKWSTPPFDPRFPSQNQIRNCYQNFLDYHRCLKTRTRRGKSTQPCEYYFRVYHSLCPISWVESWNEQIKNGIFAGKIEspecificaciones
NP_653214.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COXVIB2/MGC119094 | |
COX6B2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
125965 | |
COX6B2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MLDVEAQEPPKGKWSTPPFDPRFPSQNQIRNCYQNFLDYHRCLKTRTRRGKSTQPCEYYFRVYHSLCPISWVESWNEQIKNGIFAGKI | |
RUO | |
COX6B2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |