missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COPB Partial ORF (NP_057535, 854 a.a. - 953 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001315-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196534

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein subunit of the coatomer complex associated with non-clathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described. [provided by RefSeq]

Sequence: TVNTNMVDLNDYLQHILKSTNMKCLTPEKALSGYCGFMAANLYARSIFGEDALANVSIEKPIHQGPDAAVTGHIRIRAKSQGMALSLGDKINLSQKKTSI
Especificaciones

Especificaciones

NP_057535
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TVNTNMVDLNDYLQHILKSTNMKCLTPEKALSGYCGFMAANLYARSIFGEDALANVSIEKPIHQGPDAAVTGHIRIRAKSQGMALSLGDKINLSQKKTSI
RUO
COPB1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1315
COPB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COPB/DKFZp761K102/FLJ10341
COPB1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales