Learn More
Abnova™ Human COL25A1 Partial ORF (NP_942014.1, 57 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084570-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
COL25A1 is a brain-specific membrane-bound collagen. Proteolytic processing releases CLAC, a soluble form of COL25A1 containing the extracellular collagen domains that associates with senile plaques in Alzheimer disease (AD; MIM 104300) brains (Osada et al., 2005 [PubMed 15615705]).[supplied by OMIM]
Sequence: NDLQARIAALESAKGAPSIHLLPDTLDHLKTMVQEKVERLLAQKSYEHMAKIRIAREAPSECNCPAEspecificaciones
NP_942014.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NDLQARIAALESAKGAPSIHLLPDTLDHLKTMVQEKVERLLAQKSYEHMAKIRIAREAPSECNCPA | |
RUO | |
COL25A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84570 | |
COL25A1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLAC/CLACP | |
COL25A1 | |
Recombinant | |
wheat germ expression system |