Learn More
Abnova™ Human CNTN4 Full-length ORF (AAH26119.1, 1 a.a. - 697 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00152330-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MEENVFWECKANGRPKPTYKWLKNGEPLLTRDRIQIEQGTLNITIVNLSDAGMYQCLAENKHGVIFSNAELSVIAVGPDFSRTLLKRVTLVKVGGEVVIECKPKASPKPVYTWKKGRDILKENERITISEDGNLRIINVTKSDAGSYTCIATNHFGTASSTGNLVVKDPTRVMVPPSSMDVTVGESIVLPCQVTHDHSLDIVFTWSFNGHLIDFDRDGDHFERVGGDSAGDLMIRNIQLKHAGKYVCMVQTSVDRLSAAADLIVRGPPGPPEAVTIDEITDTTAQLSWRPGPDNHSPITMYVIQARTPFSVGWQAVSTVPELIDGKTFTATVVGLNPWVEYEFRTVAANVIGIGEPSRPSEKRRTEEALPEVTPANVSGGGGSKSELVITWETVPEELQNGRGFGYVVAFRPYGKMIWMLTVLASADASRYVFRNESVHPFSPFEVKVGVFNNKGEGPFSPTTVVYSAEEEPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRKPPPSQPPGNIIWNSSDSKIILNWDQVKALDNESEVKGYKVLYRWNRQSSTSVIETNKTSVELSLPFDEDYIIEIKPFSDGGDGSSSEQIRIPKISNAYARGSGASTSNACTLSAISTIMISLTARSSLEspecificaciones
AAH26119.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AXCAM/BIG-2/CNTN4A/MGC33615 | |
CNTN4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
152330 | |
CNTN4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEENVFWECKANGRPKPTYKWLKNGEPLLTRDRIQIEQGTLNITIVNLSDAGMYQCLAENKHGVIFSNAELSVIAVGPDFSRTLLKRVTLVKVGGEVVIECKPKASPKPVYTWKKGRDILKENERITISEDGNLRIINVTKSDAGSYTCIATNHFGTASSTGNLVVKDPTRVMVPPSSMDVTVGESIVLPCQVTHDHSLDIVFTWSFNGHLIDFDRDGDHFERVGGDSAGDLMIRNIQLKHAGKYVCMVQTSVDRLSAAADLIVRGPPGPPEAVTIDEITDTTAQLSWRPGPDNHSPITMYVIQARTPFSVGWQAVSTVPELIDGKTFTATVVGLNPWVEYEFRTVAANVIGIGEPSRPSEKRRTEEALPEVTPANVSGGGGSKSELVITWETVPEELQNGRGFGYVVAFRPYGKMIWMLTVLASADASRYVFRNESVHPFSPFEVKVGVFNNKGEGPFSPTTVVYSAEEEPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRKPPPSQPPGNIIWNSSDSKIILNWDQVKALDNESEVKGYKVLYRWNRQSSTSVIETNKTSVELSLPFDEDYIIEIKPFSDGGDGSSSEQIRIPKISNAYARGSGASTSNACTLSAISTIMISLTARSSL | |
RUO | |
CNTN4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |