missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CNAP1 Partial ORF (AAH28182, 1240 a.a. - 1339 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009918-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: LRKMLDNFDCFGDKLSDESIFSAFLSVVGKLRRGAKPEGKAIIDEFEQKLRACHTRGLDGIKELEIGQAGSQRAPSAKKPSSGSRYQPLASTASDNDFVTEspecificaciones
AAH28182 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LRKMLDNFDCFGDKLSDESIFSAFLSVVGKLRRGAKPEGKAIIDEFEQKLRACHTRGLDGIKELEIGQAGSQRAPSAKKPSSGSRYQPLASTASDNDFVT | |
RUO | |
NCAPD2 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
9918 | |
CNAP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAP-D2|CNAP1|KIAA0159|hCAP-D2 | |
NCAPD2 | |
Yes |