missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMTM6 Partial ORF (NP_060271.1, 1 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00054918-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16107796

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq]

Sequence: MENGAVYSPTTEEDPGPARGPRSGLAAYFFMGRLPLLRRVLKGLQLLLSLLAFICEEVVSQCTLCGGLYFFEFVSCSAFLLSLLI
Especificaciones

Especificaciones

NP_060271.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.98kDa
12.5% SDS-PAGE Stained with Coomassie Blue
MENGAVYSPTTEEDPGPARGPRSGLAAYFFMGRLPLLRRVLKGLQLLLSLLAFICEEVVSQCTLCGGLYFFEFVSCSAFLLSLLI
RUO
CMTM6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
54918
CMTM6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKLFSF6/FLJ20396/PRO2219
CMTM6
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales