missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLN6 Full-length ORF (NP_060352.1, 1 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00054982-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191003

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function. [provided by RefSeq]

Sequence: MEATRRRQHLGATGGPGAQLGASFLQARHGSVSADEAARTAPFHLDLWFYFTLQNWVLDFGRPIAMLVFPLEWFPLNKPSVGDYFHMAYNVITPFLLLKLIERSPRTLPRSITYVSIIIFIMGASIHLVGDSVNHRLLFSGYQHHLSVRENPIIKNLKPETLIDSFELLYYYDEYLGHCMWYIPFFLILFMYFSGCFTASKAESLIPGPALLLVAPSGLYYWYLVTEGQIFILFIFTFFAMLALVLHQKRKRLFLDSNGLFLFSSFALTLLLVALWVAWLWNDPVLRKKYPGVIYVPEPWAFYTLHVSSRH
Especificaciones

Especificaciones

NP_060352.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20561/HsT18960/nclf
CLN6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54982
CLN6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEATRRRQHLGATGGPGAQLGASFLQARHGSVSADEAARTAPFHLDLWFYFTLQNWVLDFGRPIAMLVFPLEWFPLNKPSVGDYFHMAYNVITPFLLLKLIERSPRTLPRSITYVSIIIFIMGASIHLVGDSVNHRLLFSGYQHHLSVRENPIIKNLKPETLIDSFELLYYYDEYLGHCMWYIPFFLILFMYFSGCFTASKAESLIPGPALLLVAPSGLYYWYLVTEGQIFILFIFTFFAMLALVLHQKRKRLFLDSNGLFLFSSFALTLLLVALWVAWLWNDPVLRKKYPGVIYVPEPWAFYTLHVSSRH
RUO
CLN6
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales