Learn More
Abnova™ Human CLIP2 Full-length ORF (AAI48703.1, 1 a.a. - 1046 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007461-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]
Sequence: MQKPSGLKPPGRGGKHSSPMGRTSTGSASSSAAVAASSKEGSPLHKQSSGPSSSPAAAAAPEKPGPKAAEVGDDFLGDFVVGERVWVNGVKPGVVQYLGETQFAPGQWAGVVLDDPVGKNDGAVGGVRYFECPALQGIFTRPSKLTRQPTAEGSGSDAHSVESLTAQNLSLHSGTATPPLTSRVIPLRESVLNSSVKTGNESGSNLSDSGSVKRGEKDLRLGDRVLVGGTKTGVVRYVGETDFAKGEWCGVELDEPLGKNDGAVAGTRYFQCPPKFGLFAPIHKVIRIGFPSTSPAKAKKTKRMAMGVSALTHSPSSSSISSVSSVASSVGGRPSRSGLLTETSSRYARKISGTTALQEALKEKQQHIEQLLAERDLERAEVAKATSHICEVEKEIALLKAQHEQYVAEAEEKLQRARLLVESVRKEKVDLSNQLEEERRKVEDLQFRVEEESITKGDLETQTQLEHARIGELEQSLLLEKAQAERLLRELADNRLTTVAEKSRVLQLEEELTLRRGEIEELQQCLLHSGPPPPDHPDAAEILRLRERLLSASKEHQRESGVLRDKYEKALKAYQAEVDKLRAANEKYAQEVAGLKDKVQQATSENMGLMDNWKSKLDSLASDHQKSLEDLKATLNSGPGAQQKEIGELKAVMEGIKMEHQLELGNLQAKHDLETAMHVKEKEALREKLQEAQEELAGLQRHWRAQLEVQASQHRLELQEAQDQRRDAELRVHELEKLDVEYRGQAQAIEFLKEQISLAEKKMLDYERLQRAEAQGKQEVESLREKLLVAENRLQAVEALCSSQHTHMIESNDISEETIRTKETVEGLQDKLNKRDKEVTALTSQTEMLRAQVSALESKCKSGEKKVDALLKEKRRLEAELETVSRKTHDASGQLVLISQELLRKERSLNELRVLLLEANRHSPGPERDLSREVHKAEWRIKEQKLKDDIRGLREKLTGLDKEKSLSDQRRYSLIDPSSAPELLRLQHQLMSTEDALRDALDQAQQVEKLMEAMRSCPDKAQTIGNSGSANGIHQQDKAQKQEDKHEspecificaciones
AAI48703.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
142.01kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLIP/CLIP-115/CYLN2/KIAA0291/MGC11333/WBSCR3/WBSCR4/WSCR3/WSCR4 | |
CLIP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7461 | |
CLIP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQKPSGLKPPGRGGKHSSPMGRTSTGSASSSAAVAASSKEGSPLHKQSSGPSSSPAAAAAPEKPGPKAAEVGDDFLGDFVVGERVWVNGVKPGVVQYLGETQFAPGQWAGVVLDDPVGKNDGAVGGVRYFECPALQGIFTRPSKLTRQPTAEGSGSDAHSVESLTAQNLSLHSGTATPPLTSRVIPLRESVLNSSVKTGNESGSNLSDSGSVKRGEKDLRLGDRVLVGGTKTGVVRYVGETDFAKGEWCGVELDEPLGKNDGAVAGTRYFQCPPKFGLFAPIHKVIRIGFPSTSPAKAKKTKRMAMGVSALTHSPSSSSISSVSSVASSVGGRPSRSGLLTETSSRYARKISGTTALQEALKEKQQHIEQLLAERDLERAEVAKATSHICEVEKEIALLKAQHEQYVAEAEEKLQRARLLVESVRKEKVDLSNQLEEERRKVEDLQFRVEEESITKGDLETQTQLEHARIGELEQSLLLEKAQAERLLRELADNRLTTVAEKSRVLQLEEELTLRRGEIEELQQCLLHSGPPPPDHPDAAEILRLRERLLSASKEHQRESGVLRDKYEKALKAYQAEVDKLRAANEKYAQEVAGLKDKVQQATSENMGLMDNWKSKLDSLASDHQKSLEDLKATLNSGPGAQQKEIGELKAVMEGIKMEHQLELGNLQAKHDLETAMHVKEKEALREKLQEAQEELAGLQRHWRAQLEVQASQHRLELQEAQDQRRDAELRVHELEKLDVEYRGQAQAIEFLKEQISLAEKKMLDYERLQRAEAQGKQEVESLREKLLVAENRLQAVEALCSSQHTHMIESNDISEETIRTKETVEGLQDKLNKRDKEVTALTSQTEMLRAQVSALESKCKSGEKKVDALLKEKRRLEAELETVSRKTHDASGQLVLISQELLRKERSLNELRVLLLEANRHSPGPERDLSREVHKAEWRIKEQKLKDDIRGLREKLTGLDKEKSLSDQRRYSLIDPSSAPELLRLQHQLMSTEDALRDA | |
RUO | |
CLIP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |