missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN11 Partial ORF (NP_005593, 28 a.a. - 82 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005010-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16102255

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene belongs to the claudin family of tight junction associated proteins and is a major component of central nervous system myelin that is necessary for normal CNS function. There is growing evidence that the protein determines the permeability between layers of myelin sheaths via focal adhesion and, with its expression highly regulated during development, may play an important role in cellular proliferation and migration. In addition, the protein is a candidate autoantigen in the development of autoimmune demyelinating disease. [provided by RefSeq]

Sequence: NDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACR
Especificaciones

Especificaciones

NP_005593
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.79kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACR
RUO
CLDN11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5010
CLDN11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OSP/OTM
CLDN11
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales